Learn More
CD42c Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 55161-1-AP-20UL
 View more versions of this product
Product Code. 16896635
  • 145.00 € / 20µL
Please to purchase this item. Need a web account? Register with us today!
Description
Description
The CD42c antibody from Proteintech is a rabbit polyclonal antibody to a peptide of human CD42c. This antibody recognizes human antigen. The CD42c antibody has been validated for the following applications: WB,ELISA analysis.
Specifications
Specifications
CD42c
Polyclonal
Unconjugated
20 μL
2812
Liquid
Western Blot, ELISA
180 μg/mL