Learn More
CD42c Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55161-1-AP-20UL
Description
The CD42c antibody from Proteintech is a rabbit polyclonal antibody to a peptide of human CD42c. This antibody recognizes human antigen. The CD42c antibody has been validated for the following applications: WB,ELISA analysis.
Specifications
| CD42c | |
| Polyclonal | |
| Unconjugated | |
| 20 μL | |
| 2812 | |
| Liquid |
| Western Blot, ELISA | |
| 180 μg/mL | |
| Rabbit | |
| Primary | |
| Human |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.