Learn More
CD40 Rat anti-Mouse, APC, Clone: 1C10, Proteintech
Rat Monoclonal Antibody
Brand:  Proteintech APC-65062-100UG
Product Code. 16830466
  • 171.00 € / 100µg
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
The CD40 antibody from Proteintech is a rat monoclonal antibody generated with scd40 of mouse CD40. This antibody recognizes mouse antigen. The CD40 antibody has been validated for the following applications: FC analysis. This antibody is apc.
Specifications
Specifications
CD40
Monoclonal
0.1 mg/mL
Rat
Primary
Mouse
Flow Cytometry
1C10
APC
100 μg
21939
IgG2a