Learn More
CD4 Rat anti-Mouse, FITC, Clone: GK1.5, Proteintech
Rat Monoclonal Antibody
Brand: Proteintech FITC-65104-100UG
Description
The CD4 antibody from Proteintech is a rat monoclonal antibody generated with mouse ctl clone v4 of mouse CD4. This antibody recognizes mouse antigen. The CD4 antibody has been validated for the following applications: FC analysis. This antibody is fitc.
Specifications
| CD4 | |
| Monoclonal | |
| 0.5 mg/mL | |
| Rat | |
| Primary | |
| Mouse |
| Flow Cytometry | |
| GK1.5 | |
| FITC | |
| 100 μg | |
| 12504 | |
| IgG2b |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.