Learn More
CD3e Hamster, anti-Mouse, PE, Clone: 145-2C11, STEMCELL Technologies
Hamster Monoclonal Antibody
Brand: Stemcell Technologies 60015PE.1
Description
- The 145-2C11 antibody reacts with the ∼20 kDa CD3ε subunit of the mouse T cell receptor (TCR)/CD3 complex, which is expressed on the surface of circulating mature T cells and NKT cells, and variably on thymocytes
- A majority of T cell neoplasms also express CD3
- Activation of T cells by the TCR involves the cytoplasmic tails of the CD3 subunits, which are structurally related type 1 transmembrane proteins and members of the immunoglobulin superfamily
- The 145-2C11 antibody has been used for in vitro functional (blocking and activation) assays and has been reported to block binding by the clone 17A2 antibody
- The 145-2C11 antibody is not recommended for use with formalin-fixed, paraffin-embedded sections
- This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Mouse T Cell Isolation Kit (Catalog #19851) and EasySep™ Mouse CD90.2 Positive Selection Kit II (Catalog #18951)
Specifications
| CD3e | |
| Monoclonal | |
| PE | |
| 50 μg | |
| Mouse | |
| T Cells |
| Immunohistochemistry, Western Blot, ELISA, Flow Cytometry, Immunocytochemistry | |
| 145-2C11 | |
| Hamster | |
| Primary | |
| IgG1 κ |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.