Learn More
CD3e Hamster, anti-Mouse, PE, Clone: 145-2C11, STEMCELL Technologies
Hamster Monoclonal Antibody
Brand:  Stemcell Technologies 60015PE.1
Encompass_Preferred
 View more versions of this product
Product Code. 17118461
  • 135.00 € / 50µg
Please to purchase this item. Need a web account? Register with us today!
Description
Description
  • The 145-2C11 antibody reacts with the ∼20 kDa CD3ε subunit of the mouse T cell receptor (TCR)/CD3 complex, which is expressed on the surface of circulating mature T cells and NKT cells, and variably on thymocytes
  • A majority of T cell neoplasms also express CD3
  • Activation of T cells by the TCR involves the cytoplasmic tails of the CD3 subunits, which are structurally related type 1 transmembrane proteins and members of the immunoglobulin superfamily
  • The 145-2C11 antibody has been used for in vitro functional (blocking and activation) assays and has been reported to block binding by the clone 17A2 antibody
  • The 145-2C11 antibody is not recommended for use with formalin-fixed, paraffin-embedded sections
  • This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Mouse T Cell Isolation Kit (Catalog #19851) and EasySep™ Mouse CD90.2 Positive Selection Kit II (Catalog #18951)
Specifications
Specifications
CD3e
Monoclonal
PE
50 μg
Mouse
T Cells
Immunohistochemistry, Western Blot, ELISA, Flow Cytometry, Immunocytochemistry
145-2C11
Hamster
Primary
IgG1 κ