missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CD36 Peptide P (93-110)

Product Code. 15418559
Change view
Click to view available options
Quantity:
0.5 mg
1 mg
Unit Size:
0.5mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15418559 1 mg 1mg
15408559 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15418559 Supplier Thermo Scientific™ Supplier No. J66970.M^
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For cellular and molecular biology applications

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

Purification Method HPLC
Content And Storage -30°C to -10°C
Format Lyophilized Powder
Molecular Weight (g/mol) 2270.45
Product Type CD36 Peptide P (93-110)
Quantity 1 mg
Regulatory Status RUO
Sequence Tyr-Arg-Val-Arg-Phe-Leu-Ala-Lys-Glu-Asn-Val-Thr-Gln-Asp-Ala-Glu-Asp-Asn-Cys

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.