missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD357 (AITR/GITR) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA546810
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: LRSQCMWPRE TQLLLEVPPS TEDARSCQFP EEERGERSAE EKGRLGDLWV Sequence homology: Human: 100%
CD357 (AITR/GITR) Polyclonal antibody specifically detects CD357 (AITR/GITR) in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
CD357 (AITR/GITR) | |
Polyclonal | |
Unconjugated | |
Tnfrsf18 | |
Activation-inducible TNFR family receptor; AITR; CD357; GITR; GITR-D; glucocorticoid-induced TNFR-related protein; PRO364; TNF receptor superfamily activation-inducible protein; TNF receptor superfamily member 18; TNFRSF18; Tumor necrosis factor receptor superfamily member 18; tumor necrosis factor receptor superfamily, member 18; UNQ319; UNQ319/PRO364 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
8784 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9Y5U5 | |
Tnfrsf18 | |
Synthetic peptide directed towards the C-terminal of human TNFRSF18 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |