missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD34 Rat anti-Mouse, BV786, Clone: RAM34, BD Biosciences™
Rat Monoclonal Antibody
Brand: BD Biosciences 742971
This item is not returnable.
View return policy
Description
CD34 Monoclonal antibody specifically detects CD34 in Mouse samples. It is validated for Flow Cytometry
Specifications
| CD34 | |
| Monoclonal | |
| Brilliant Violet 786 | |
| Cd34; CD34 antigen; Hematopoietic progenitor cell antigen CD34 | |
| Recombinant Mouse CD34 | |
| RUO | |
| Mouse | |
| IgG2a κ |
| Flow Cytometry | |
| RAM34 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Rat | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction