Learn More
Invitrogen™ CD321 (F11R) Monoclonal Antibody (CSTEM27), Alexa Fluor™ 488, eBioscience™, Invitrogen™
Description
This monoclonal antibody CSTEM27 reacts with human CD321 (F11R, JAM-1, JAM-A). Applications Reported: This CSTEM27 antibody has been reported for use in immunocytochemical staining and flow cytometric analysis. This CSTEM27 antibody has been tested by immunocytochemistry of adherent cells. This can be used at less than or equal to 5 μg/mL. This CSTEM27 antibody has also been tested by flow cytometric analysis of human iPSCs. This may be used at 0.25 μg/test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.
Specifications
Specifications
| Antigen | CD321 (F11R) |
| Applications | Flow Cytometry, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | CSTEM27 |
| Concentration | 5 μL/Test |
| Conjugate | Alexa Fluor 488 |
| Formulation | PBS with BSA and 0.09% sodium azide; pH 7.2 |
| Gene | F11r |
| Gene Accession No. | Q9Y624 |
| Gene Alias | 9130004G24; AA638916; BV11 antigen; CD321; ESTM33; F11 R; F11 receptor; F11 receptor/F11R; F11R; JAM; JAM1; JAM-1; JAMA; JAM-A; Jcam; Jcam1; junction cell adhesion molecule A; junction cell adhesion molecule1; junctional adhesion molecule 1; Junctional adhesion molecule A; KAT; Ly106; PAM-1; Platelet adhesion molecule 1; platelet F11 receptor; UNQ264/PRO301 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.