Learn More
CD32 Mouse anti-Rat, Unlabeled, Clone: D34-485, BD
Description
The D34-485 antibody reacts specifically with CD32, the FcγII receptor. Rat CD32 is expressed on B lymphocytes, myeloid cells, and some lymphocytes in the thymic medulla. D34-485 mAb blocks binding of aggregated immunoglobulins to the FcγIi receptors in vitro. This antibody is routinely tested by flow cytometric analysis and in vitro blocking. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Mouse
Clone: D34-485
Isotype: IgG1 κ
Species Reactivity: Rat
Immunogen: Recombinant Rat CD32 Protein
Blocking Assay, Flow Cytometry
Specifications
Specifications
| Antigen | CD32 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | D34-485 |
| Concentration | 0.5mg/mL |
| Conjugate | Unconjugated |
| Description | Fcgr2b; FcRII; Fc-gamma RII; FcγRII; FcγII receptor |
| Formulation | Aqueous buffered solution containing ≤0.09% sodium azide. |
| Host Species | Mouse |
| Immunogen | Recombinant Rat CD32 Protein |
| Show More |
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.