missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD300f (IREM-1) Monoclonal Antibody (UP-D1), eFluor™ 660, eBioscience™
GREENER_CHOICE

Product Code. 15539926
Change view
Click to view available options
Quantity:
100 Tests
Unit Size:
100 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15539926 100 Tests 100 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15539926 Supplier Invitrogen™ Supplier No. 50300842

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Description: The monoclonal antibody recognizes human IREM-1 (Immune receptor expressed by myeloid cell 1). IREM-1 is also a member of of the CD300 family specifically referred to as CD300f. IREM-1 is a type I transmembrane protein with a single Ig domain, one predicted N-linked glycosylation site and a long cytoplasmic tail which contains a predicted ITIM domain categorizing it as an inhibitory receptor. Additionally the cytoplasmic domain has been shown to interact with SHP-1 and the p85\alpha subunit of PI3K. Expression is restricted to myeloid cells. In peripheral blood IREM-1 is found on monocytes and granulocytes. In monocyte derived dendritic cells, expression of IREM-1 is downregulated. Applications Reported: This UP-D1 antibody has been reported for use in flow cytometric analysis. Applications Tested: This UP-D1 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. eFluor® 660 is a replacement for Alexa Fluor® 647. eFluor® 660 emits at 659 nm and is excited with the red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome. Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser. Filtration: 0.2 ...

CLM1 is an inhibitory receptor of the Ig superfamily expressed on myeloid cells. It mediates negative regulatory signals by recruiting SHP1 (PTPN6; MIM 176883) or SHIP.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD300f (IREM-1)
Applications Flow Cytometry
Classification Monoclonal
Clone UP-D1
Concentration 5 μL/Test
Conjugate eFluor 660
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene CD300LF
Gene Accession No. Q8TDQ1
Gene Alias CD300 antigen like family member F; CD300 antigen-like family member F; CD300 molecule like family member f; CD300 molecule-like family member f; CD300f; Cd300lf; CLIM1; CLM1; CLM-1; CMRF-35-like molecule 1; CMRF35-like molecule 1; DC-derived Ig-like receptor 2; Digr2; F730004D16Rik; IgSF13; Immune receptor expressed on myeloid cells 1; immunoglobin superfamily member 13; immunoglobulin superfamily member 13; inhibitory receptor IREM1; IREM1; IREM-1; Leukocyte mono-Ig-like receptor 3; Lmir3; MAIR-5; MAIR-V; myeloid-associated immunoglobulin-like receptor 5; NK inhibitory receptor; NKIR; Pigr3; UNQ3105/PRO10111
Gene Symbols CD300LF
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 146722
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.