Learn More
CD30/TNFRSF8 Mouse anti-Human, Alexa Fluor 700, Clone: 81337, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB229N-100UG
Description
CD30/TNFRSF8 Monoclonal specifically detects CD30/TNFRSF8 in Human samples. It is validated for Flow Cytometry.Specifications
| CD30/TNFRSF8 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| P28908 | |
| TNFRSF8 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human CD30/TNFRSF8 Phe19-Lys379 Accession # P28908 | |
| RUO | |
| 943 | |
| Human | |
| IgG2b |
| Flow Cytometry | |
| 81337 | |
| Alexa Fluor 700 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| CD30, CD30 antigen, CD30KI-1, CD30L receptor, cytokine receptor CD30, D1S166EKi-1, Ki-1 antigen, Lymphocyte activation antigen CD30, TNFRSF8, tumor necrosis factor receptor superfamily member 8, tumor necrosis factor receptor superfamily, member 8 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human CD30/TNFRSF8 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant mouse CD30, recombinant human (rh) CD27, and rhCD40 is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
For Research Use Only (RUO)