missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD30/TNFRSF8 Antibody (rKi-1/779), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-54340AF700
This item is not returnable.
View return policy
Description
CD30/TNFRSF8 Monoclonal antibody specifically detects CD30/TNFRSF8 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF
Specifications
| CD30/TNFRSF8 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| CD30, CD30 antigen, CD30KI-1, CD30L receptor, cytokine receptor CD30, D1S166EKi-1, Ki-1 antigen, Lymphocyte activation antigen CD30, tumor necrosis factor receptor superfamily member 8, tumor necrosis factor receptor superfamily, member 8 | |
| Recombinant full-length human CD30/TNFRSF8 protein (Uniprot: P28908) | |
| 100 μg | |
| Apoptosis, B Cell Development and Differentiation Markers, Cell Cycle and Replication, Immunology, Stem Cell Markers | |
| 943 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF | |
| rKi-1/779 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction