missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD3 Mouse, anti-Human, FITC, Clone: OKT3, STEMCELL Technologies
Description
- The OKT3 antibody reacts with an extracellular conformational epitope on the ∼20 kDa CD3ε subunit of the human T cell receptor (TCR)/CD3 complex, which is expressed on the surface of ∼95% of mature T cells and NKT cells, and variably on thymocytes
- A majority of T cell neoplasms also express CD3
- CD3 is a dimer assembled from combinations of CD3γ, δ, ε, η, and ζ subunits, which are structurally related type 1 transmembrane proteins and members of the immunoglobulin superfamily
- CD3 associates non-covalently with the TCR and is involved in transducing antigen recognition signals into the cytoplasm of T cells and in regulating T cell activation and the expression of the TCR on the cell surface
- Mutations in CD3 have been associated with disorders of the immune system, including severe combined immunodeficiency (SCID)
- The OKT3 antibody recognizes an epitope that is expressed when the CD3ε subunit associates with either CD3γ or CD3δ, and binding of the antibody can induce T cell activation
- Because of its immunosuppressive properties in vivo, the OKT3 antibody has proven useful in treating allograft rejection
- Clone OKT3 is widely used for phenotyping human T cells, and it has been observed to block the binding of anti-CD3 antibody clones SK7 and UCHT1
Specifications
Specifications
| Antigen | CD3 |
| Applications | Immunohistochemistry, Western Blot, Flow Cytometry, Functional Assay |
| Classification | Monoclonal |
| Clone | OKT3 |
| Conjugate | FITC |
| Host Species | Mouse |
| Quantity | 25 Tests |
| Primary or Secondary | Primary |
| Target Species | Human |
| Isotype | IgG2a κ |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?