Learn More
CD3 Mouse anti-Human, CoraLite 647, Clone: OKT3, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech CL647-65133-100TESTS
Product Code. 16801146
  • 135.00 € / 100 tests
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
The CD3 antibody from Proteintech is a mouse monoclonal antibody generated with n/a of human CD3. This antibody recognizes human antigen. The CD3 antibody has been validated for the following applications: FC analysis. This antibody is coralite™647.
Specifications
Specifications
CD3
Monoclonal
5 μL/Test
Mouse
Primary
Human
Flow Cytometry
OKT3
CoraLite 647
100 Tests
916
IgG2a