missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD297 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA550607
Additional Details : Weight : 0.01000kg
Description
The antibody detects endogenous levels of total CD297 / ART4 protein.
This gene encodes a protein that contains a mono-ADP-ribosylation motif. It is a member of the ADP-ribosyltransferase gene family but enzymatic activity has not been demonstrated experimentally. Antigens of the Dombrock blood group system are located on the gene product, which is glycosylphosphatidylinosotol-anchored to the erythrocyte membrane. Allelic variants, some of which lead to adverse transfusion reactions, are known.Specifications
CD297 | |
Polyclonal | |
Unconjugated | |
Art4 | |
4432404K01Rik; ADP-ribosyltransferase 4; ADP-ribosyltransferase 4 (DO blood group); ADP-ribosyltransferase 4 (Dombrock blood group); ADP-ribosyltransferase 4 (inactive) (Dombrock blood group); ADP-ribosyltransferase C2 and C3 toxin-like 4; ART4; ARTC4; ATR4; CD297; DO; DOK1; Dombrock blood group carrier molecule; ecto-ADP-ribosyltransferase 4; mono (ADP-ribosyl)transferase; mono(ADP-ribosyl)transferase 4; mono-ADP-ribosyltransferase 4; NAD(+)--protein-arginine ADP-ribosyltransferase 4; NAD(P)(+)--arginine ADP-ribosyltransferase; NAD(P)(+)--arginine ADP-ribosyltransferase 4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
109978, 420 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
2 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q93070, Q9CRA0 | |
Art4 | |
Synthetic peptide corresponding to a region derived from internal residues of human ADP-ribosyltransferase 4 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |