missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD28 Antibody (PV-1) - Azide and BSA Free, Novus Biologicals™
Armenian Hamster Monoclonal Antibody
Brand: Novus Biologicals NBP1-27932
This item is not returnable.
View return policy
Description
CD28 Monoclonal specifically detects CD28 in Mouse samples. It is validated for Flow Cytometry.
Specifications
| CD28 | |
| Monoclonal | |
| 0.5 mg/mL | |
| Flow Cytometry 1:10 - 1:1000 | |
| CD28 | |
| C57BL/6N mouse T cell lymphoma EL4 cell line | |
| 0.5 mg | |
| Adaptive Immunity, Apoptosis, Cancer, Cell Biology, Immune System Diseases, Immunology, Neuroscience, Signal Transduction | |
| 940 | |
| Mouse | |
| Purified |
| Flow Cytometry, Immunoprecipitation, In vitro Assay | |
| PV-1 | |
| Unconjugated | |
| CD28 antigen, CD28 antigen (Tp44), CD28 molecule, MGC138290, T-cell-specific surface glycoprotein CD28, Tp44 | |
| Armenian Hamster | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Mouse CD28 | |
| Store at 4C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction