Learn More
Novus Biologicals™ CD28 Antibody (FR104) - Humanized, IgG2SA, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-28252-50ug
 View more versions of this product
Product Code. 30044189
  • 213.00 € / 50µg
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
CD28 Monoclonal antibody specifically detects CD28 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD28
Monoclonal
Unconjugated
P10747
Human
Protein A purified
RUO
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.
IgG2
Flow Cytometry, ELISA, Functional Assay
FR104
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
CD28 antigen, CD28 antigen (Tp44), CD28 molecule, MGC138290, T-cell-specific surface glycoprotein CD28, Tp44
CD28
50 μg
Adaptive Immunity, Apoptosis, Cancer, Cell Biology, Immune System Diseases, Immunology, Neuroscience, Signal Transduction
940
Human
Purified