Learn More
CD279 (PD-1) Mouse anti-Human, APC, Clone: J110, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech APC-65119-100TESTS
Description
The CD279 (PD-1) antibody from Proteintech is a mouse monoclonal antibody generated with n/a of human CD279 (PD-1). This antibody recognizes human antigen. The CD279 (PD-1) antibody has been validated for the following applications: FC analysis. This antibody is apc.
Specifications
| CD279 (PD-1) | |
| Monoclonal | |
| 5 μL (1 μg)/Test | |
| Mouse | |
| Primary | |
| Human |
| Flow Cytometry | |
| J110 | |
| APC | |
| 100 Tests | |
| 5133 | |
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.