missing translation for 'onlineSavingsMsg'
Learn More

PeproTech CD267 (TACI) Polyclonal Antibody, PeproTech®, Invitrogen™
GREENER_CHOICE

Product Code. 17834383
Change view
Click to view available options
Quantity:
1 mg
100 μg
50 μg
Unit Size:
100µg
1mg
50µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17834383 100 μg 100µg
17804173 50 μg 50µg
17813993 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17834383 Supplier PeproTech Supplier No. 500P166G100UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

AA Sequence of recombinant protein: SGLGRSRRGG RSRVDQEERF PQGLWTGVAM RSCPEEQYWD PLLGTCMSCK TICNHQSQRT CAAFCRSLSC RKEQGKFYDH LLRDCISCAS ICGQHPKQCA YFCENKLRSP VNLPPELRRQ RSGEVENNSD NSGRYQGLEH RGSEASPALP GLKLSADQV. Preparation: Produced from sera of goats immunized with highly pure Recombinant Human TACI. Anti-Human TACI-specific antibody was purified by affinity chromatography employing an immobilized Human TACI matrix. Sandwich ELISA: To detect Human TACI by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human TACI (500-P166GBt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human TACI. Western Blot: To detect Human TACI by Western Blot analysis this antibody can be used at a concentration of 0.1- 0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Human TACI is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.

Members in the TNF superfamily regulate immune responses and induce apoptosis. Two novel members in the TNF family were recently identified and designated BAFF/BLyS/TALL-1/THANK/zTNF4 and April/TALL-2, respectively. BAFF was characterized as a B cell activator since it induced B cell proliferation and immunoglobulin secretion. April regulates immunological and non-immunological cell growth. Three receptors, BCMA, TACI, and BAFF-R, for BAFF and April were recently identified. TACI, like BCMA, binds BAFF and April. TACI and its ligands regulate humoral immune responses, activate NF-kappa-B and c-jun N-terminal kinase, and are involved in B cell associated autoimmune diseases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD267 (TACI)
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.1-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene TNFRSF13B
Gene Accession No. O14836
Gene Alias 1200009E08Rik; CD267; CVID; CVID2; IGAD2; NEWGENE_1595998; RYZN; TACI; TNF receptor superfamily member 13B; Tnfrsf13b; TNFRSF14B; transmembrane activator and calcium-modulating and cyclophilin ligand interactor; transmembrane activator and CAML interactor; tumor necrosis factor receptor 13B; tumor necrosis factor receptor superfamily member 13B; tumor necrosis factor receptor superfamily, member 13B
Gene Symbols TNFRSF13B
Host Species Goat
Immunogen E.coli-derived Recombinant Human TACI.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23495
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.