Learn More
Invitrogen™ CD218a (IL-18Ra) Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5115404
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total IL18R1.
IL18R1 is a cytokine receptor that belongs to the interleukin 1 receptor family. This receptor specifically binds interleukin 18 (IL18), and is essential for IL18 mediated signal transduction. IFN-alpha and IL12 are reported to induce the expression of this receptor in NK and T cells. The gene along with four other members of the interleukin 1 receptor family, including IL1R2, IL1R1, ILRL2 (IL-1Rrp2), and IL1RL1 (T1/ST2), form a gene cluster on chromosome 2q.
Specifications
| CD218a (IL-18Ra) | |
| Polyclonal | |
| Unconjugated | |
| Il18r1 | |
| CD218 antigen-like family member A; CD218a; CDw218a; cytokine receptor; IL1 receptor-related protein; Il18r1; IL-18R1; IL-18R-1; IL18RA; Il18ralpha; IL-18R-alpha; IL18Ralpha2; IL1RRP; IL-1Rrp; IL1R-rp; interleukin 1 receptor related protein; interleukin 18 receptor 1; Interleukin-18; interleukin-18 receptor 1; Interleukin-18 receptor alpha | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 8809 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| Q13478 | |
| Il18r1 | |
| A synthesized peptide derived from human IL18R1(Accession Q13478), corresponding to amino acid residues V233-R283. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |