Learn More
Invitrogen™ CD218a (IL-18Ra) Monoclonal Antibody (H44), APC, eBioscience™

Mouse Monoclonal Antibody
384.00 €
Specifications
| Antigen | CD218a (IL-18Ra) |
|---|---|
| Clone | H44 |
| Concentration | 5 μL/Test |
| Content And Storage | 4°C, store in dark, DO NOT FREEZE! |
| Applications | Flow Cytometry |
Description
Description: The H44 antibody reacts with the interleukin-18 receptor alpha chain, also known as IL-1Rrp, an IL-1R family member. IL-18R alpha is expressed on Th1 cells, NK cell subset, neutrophils and IL-12-stimulated tonsillar B cells. By itself, IL-18R alpha binds IL-18 with low affinity and when complexed with IL-18R beta chain binds IL-18 with high affinity. The proinflammatory cytokine IL-18 and its receptor complex play an important role in the innate and autoimmune responses. Applications Reported: This H44 antibody has been reported for use in flow cytometric analysis. Applications Tested: This H44 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended to stain whole blood before RBC lysis. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.
IL18R1 is a cytokine receptor that belongs to the interleukin 1 receptor family. This receptor specifically binds interleukin 18 (IL18), and is essential for IL18 mediated signal transduction. IFN-alpha and IL12 are reported to induce the expression of this receptor in NK and T cells. The gene along with four other members of the interleukin 1 receptor family, including IL1R2, IL1R1, ILRL2 (IL-1Rrp2), and IL1RL1 (T1/ST2), form a gene cluster on chromosome 2q.Specifications
| CD218a (IL-18Ra) | |
| 5 μL/Test | |
| Flow Cytometry | |
| APC | |
| Mouse | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| Q13478 | |
| 8809 | |
| Primary | |
| Affinity chromatography |
| H44 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Monoclonal | |
| Liquid | |
| IgG1 κ | |
| Human | |
| Il18r1 | |
| CD218 antigen-like family member A; CD218a; CDw218a; cytokine receptor; IL1 receptor-related protein; Il18r1; IL-18R1; IL-18R-1; IL18RA; Il18ralpha; IL-18R-alpha; IL18Ralpha2; IL1RRP; IL-1Rrp; IL1R-rp; interleukin 1 receptor related protein; interleukin 18 receptor 1; Interleukin-18; interleukin-18 receptor 1; Interleukin-18 receptor alpha | |
| Il18r1 | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.