missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD203c Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA532749
This item is not returnable.
View return policy
Description
Percent identity with other species by BLAST analysis: Human, Gibbon, Monkey (100%) Orangutan, Marmoset (95%) Horse, Rabbit (84%).
The protein encoded by this gene belongs to a series of ectoenzymes that are involved in hydrolysis of extracellular nucleotides. These ectoenzymes possess ATPase and ATP pyrophosphatase activities and are type II transmembrane proteins. Expression of the related rat mRNA has been found in a subset of immature glial cells and in the alimentary tract. The corresponding rat protein has been detected in the pancreas, small intestine, colon, and liver. The human mRNA is expressed in glioma cells, prostate, and uterus. Expression of the human protein has been detected in uterus, basophils, and mast cells. [provided by RefSeq, Jul 2008].
Specifications
| CD203c | |
| Polyclonal | |
| Unconjugated | |
| ENPP3 | |
| AI876438; alkaline phosphodiesterase; Alkaline phosphodiesterase I; B10; CD203c; dJ1005H11.3 (phosphodiesterase I/nucleotide pyrophosphatase 3); dJ914N13.3 (phosphodiesterase I/nucleotide pyrophosphatase 3); ectonucleotide pyrophosphatase/phosphodiesterase 3; ectonucleotide pyrophosphatase/phosphodiesterase family member 3; E-NPP 3; ENPP3; gp130RB13-6; Npp3; NPPase; Nucleotide diphosphatase; Nucleotide pyrophosphatase; PD-Ibeta; Pdnp3; Phosphodiesterase I beta; phosphodiesterase I/nucleotide pyrophosphatase 3; phosphodiesterase-I beta; RB13-6; RB13-6 antigen | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 5169 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.6 mg/mL | |
| PBS with 0.1% sodium azide | |
| O14638 | |
| ENPP3 | |
| Synthetic 19 amino acid peptide from internal region of human ENPP3. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction