missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD20 Antibody (10F381 (rituximab)), Alexa Fluor™ 647 - Chimeric, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-52646AF647
This item is not returnable.
View return policy
Description
CD20 Monoclonal antibody specifically detects CD20 in Human, Cynomolgus Monkey, Macaque samples. It is validated for Flow Cytometry, CyTOF
Specifications
| CD20 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| B1, B-lymphocyte antigen CD20, B-lymphocyte cell-surface antigen B1, B-lymphocyte surface antigen B1, Bp35MGC3969, CD20 antigen, CD20 receptor, CD20S7, CVID5, LEU-16, Leukocyte surface antigen Leu-16, Membrane-spanning 4-domains subfamily A member 1, membrane-spanning 4-domains, subfamily A, member 1, MS4A2 | |
| Human lymphoblastoid cell line SB. | |
| 0.1 mL | |
| Adaptive Immunity, B Cell Development and Differentiation Markers, Cancer, Cancer Stem Cells, Cell Biology, Cytokine Research, Immunology, Signal Transduction, Stem Cell Markers, Tumor Biomarkers | |
| 931 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Flow Cytometry, CyTOF | |
| 10F381 (rituximab) | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Cynomolgus Monkey, Macaque | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction