missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD185 (CXCR5) Monoclonal Antibody (MU5UBEE), Brilliant Ultra Violet™ 615, eBioscience™, Invitrogen™

Product Code. 17885107
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
Unit Size:
Pack of 100
Pack of 25
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17885107 100 Tests Pack of 100
17875027 25 Tests Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17885107 Supplier Invitrogen™ Supplier No. 366918542

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD185, also known as CXCR5 or Burkitt lymphoma receptor 1 (BLR1), is a seven-transmembrane G protein-coupled receptor in the CXC chemokine receptor family. It plays a crucial role in lymphocyte trafficking, particularly in T cell migration into B cell follicles of germinal centers in response to CXCL13, making it a key marker of follicular helper T cells. In peripheral blood, CXCR5 is expressed on B cells, CD4+ T cells (excluding Th1 and Th2 cells), and a subset of memory (CD45RO+) T cells. These CD185+ T cells are in a resting state and migrate to lymph nodes due to CCR7 and CD62L expression. In tonsils, CD185 is found on nearly all CD4+ cells, along with activation markers like CD69 and ICOS, and can induce antibody production when co-cultured with B cells. CXCR5 is expressed in lymphatic tissues such as lymph node follicles and the spleen, and is also found in human blood and salivary glands, as well as various animal tissues. It is essential for B cell migration and is part of the larger family of G protein-coupled receptors (GPCRs), which have seven membrane-spanning helices and are involved in numerous signaling pathways. GPCRs, including CXCR5, are regulated by various ligands that affect their conformation and signaling through G-proteins and downstream effectors. The regulation of GPCRs, including their deactivation, has significant implications for cellular functions and is a major focus of pharmacological research, leading to the development of drugs like Zyprexa and Claritin for mental health and allergy treatments.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD185 (CXCR5)
Applications Flow Cytometry
Classification Monoclonal
Clone MU5UBEE
Concentration 5 μL/Test
Conjugate Brilliant Ultraviolet 615
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene CXCR5
Gene Accession No. P32302
Gene Alias Blr1; Blr-1; Burkitt lymphoma receptor 1; burkitt lymphoma receptor 1 homolog; Burkitt lymphoma receptor 1, GTP binding protein (chemokine (C-X-C motif) receptor 5); Burkitt lymphoma receptor 1, GTP-binding protein; C Cmotif chemokine; C X C motif chemokine; CC motif chemokine; CCmotif chemokine; CD185; CD185 antigen; chemochine (C-X-C motif) receptor 5; chemokine (C-X-C motif) receptor 5; CXC; C-X-C chemokine receptor type 5; CXC motif chemokine; C-X-C motif chemokine receptor 5; CXCR5; CXC-R5; CXCR-5; EGK_06966; Fusin; Gpcr6; GPR9; HUMSTR; LESTR; MDR15; MDR-15; Monocyte-derived receptor 15; neurolymphatic receptor; NLR
Gene Symbols CXCR5
Host Species Mouse
Immunogen N-terminal extracellular domain.
Purification Method Affinity chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 643, 701792
Target Species Human, Monkey, Rhesus Macaque
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.