missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD163 Antibody (K20-T), Novus Biologicals™
Rabbit Monoclonal Antibody has been used in 7 publications
Brand: Novus Biologicals NBP1-30148-0.1ml
This item is not returnable.
View return policy
Description
CD163 Monoclonal specifically detects CD163 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| CD163 | |
| Monoclonal | |
| 2.mg/mL | |
| Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:100-200, Immunohistochemistry-Frozen 1:10-1:500 | |
| Q86VB7 | |
| CD163 | |
| Peptide derived from N-terminal sequence of human CD163. | |
| 0.1 mL | |
| Primary | |
| This antibody is specific for the N terminus of CD163 | |
| Store at 4C. Do not freeze. |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| K20-T | |
| Unconjugated | |
| 20mM Tris/HCl (pH 8.0) and 20 mg/ml BSA with 0.05% Sodium Azide | |
| CD163 antigenscavenger receptor cysteine-rich type 1 protein M130, CD163 molecule, Hemoglobin scavenger receptor, M130, M130macrophage-associated antigen, MM130 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 9332 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction