Learn More
CD16 Mouse, anti-Human, FITC, Clone: 3G8, STEMCELL Technologies
Mouse Monoclonal Antibody
Brand:  Stemcell Technologies 60041FI
Encompass_Preferred
 View more versions of this product
Product Code. 17198731
  • 433.35 € / 100 tests
Please to purchase this item. Need a web account? Register with us today!
Description
Description
  • The 3G8 antibody reacts with CD16 (FcγRIII or low affinity IgG receptor III), a type 1 transmembrane glycoprotein belonging to the Ig superfamily
  • CD16 exists in two distinct isoforms: CD16a, a 50 - 65 kDa form expressed on NK cells, activated monocytes, macrophages, and placental trophoblasts, and CD16b, an ∼48 kDa glycosylphosphatidylinositol (GPI)-anchored form expressed on neutrophils, basophils and eosinophils and found as at least two polymorphic variants, termed NA1 and NA2
  • CD16 binds weakly to the Fc region of monomeric, aggregated or complexed IgG, particularly the IgG1 and IgG3 isotypes
  • Binding of IgG to either CD16 isoform induces signaling pathways that modulate several types of responses, including antibody-dependent cell-mediated cytotoxicity (ADCC), phagocytosis, cytokine release and proliferation
  • CD16/IgG interactions can result in non-specific labeling in antibody-based detection and cell separation experiments
Specifications
Specifications
CD16
Monoclonal
FITC
100 Tests
Baboon, Cynomolgus, Chimpanzee, Human
Granulocytes and Subsets, Monocytes
Immunohistochemistry, ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence
3G8
Mouse
Primary
IgG1 κ