missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD158a Mouse anti-Human, FITC, Clone: HP-3E4, BD
Description
CD158a, clone HP-3E4, is derived from spleen cells from BALB/c mice immunized with activated NK lymphocytes and fused with SP2 myeloma cells.
CD158a recognizes a 58-kdalton (kDa) surface protein found on natural killer (NK) cells and on some CD4– T lymphocytes.
Specifications
Specifications
| Antigen | CD158a |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | HP-3E4 |
| Conjugate | FITC |
| Formulation | Phosphate buffered saline with gelatin and 0.1% sodium azide. |
| Host Species | Mouse |
| Immunogen | Human NK Cells |
| Quantity | 50 Tests |
| Regulatory Status | RUO |
| Show More |
For Research Use Only.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?