missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD151 Antibody (11G5a), PE/Atto594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-64810PEATT594
This item is not returnable.
View return policy
Description
CD151 Monoclonal specifically detects CD151 in Human samples. It is validated for Flow Cytometry.
Specifications
| CD151 | |
| Monoclonal | |
| PE/ATTO 594 | |
| PBS with 0.05% Sodium Azide | |
| CD151 | |
| Made to Human CD151 | |
| 0.1 mL | |
| 977.0 | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| 11G5a | |
| Flow Cytometry | |
| CD151 antigen (Raph blood group), CD151 antigenplatelet surface glycoprotein gp27, CD151 molecule (Raph blood group), GP27, Membrane glycoprotein SFA-1, MER2, PETA3, PETA-3SFA-1, platelet-endothelial cell tetraspan antigen 3, Platelet-endothelial tetraspan antigen 3, RAPH, SFA1, tetraspanin-24, tspan-24, TSPAN24hemidesmosomal tetraspanin CD151 | |
| Mouse | |
| Protein G purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction