Learn More
CD124 Mouse anti-Human, BUV395, Clone: hIL4R-M57, BD Optibuild™
Mouse Monoclonal Antibody
Brand: BD Biosciences 743163
Description
The hIL4R-M57 antibody specifically binds to the α subunit (IL-4Rα) of the human Interleukin-4 Receptor complex (CD124). The human IL-4Rα, also known as B cell stimulatory factor 1 receptor (BSF-1 receptor), is a 140kDa transmembrane glycoprotein that is expressed by B and T lymphocytes and a variety of other hematopoietic and non-hematopoietic cells and cell lines. The cell surface IL-4Rα chain binds IL-4 with high affinity and associates with either the common γ chain (IL-4Rα/γ c; aka, type I IL-4R complex) or the IL-13 receptor alpha-1 subunit (IL-4Rα/IL-13Rα 1; aka, type II IL-4R complex) to form two distinct types of signal-transducing IL-4R complexes. The type I IL-4 receptor complex specifically binds IL-4 whereas the type II IL-4R complex binds and transduces signals from either IL-4 or IL-13. A truncated form of the IL-4Rα exists in soluble form in biological fluids. In contrast to mice, in humans no distinct mRNA coding for sIL-4R has been described, suggested that human sIL4-R is exclusively produced by proteolytic cleavage of the cell surface receptor. The serum levels of soluble IL-4Rα appear to elevate in pathological situations such as allergy and parasitic infections. Depending on the ratios of IL-4 and sIL-4Rα present in the local milieu, the sIL-4Rα may augment or antagonize the activities of IL-4. The immunogen used to generate the hIL4R-M57 hybridoma was soluble human IL-4R.
The antibody was conjugated to BD Horizon™ BUV395 which is part of the BD Horizon Brilliant™ Ultraviolet family of dyes. This dye has been exclusively developed by BD Biosciences to have minimal spillover into other detectors, making it an optimal choice for multicolor flow cytometry. With an Ex Max at 348 nm and an Em Max at 395 nm, BD Horizon BUV395 can be excited with a 355 nm laser and detected with a 379/28 filter. BUV395 is a UV-excitable dye that has been developed exclusively by BD Biosciences. With an excitation max of 348 nm and emission max of 395 nm, BUV395 can be excited by the 355nm laser and detected in a 379/28 filter. This dye is optimal for multicolor flow cytometry because it has little to no spillover into other detectors.
Specifications
| CD124 | |
| hIL4R-M57 | |
| Brilliant Ultraviolet 395 | |
| IL-4 Receptor α Chain, CD124 | |
| Soluble Human IL-4 Receptor | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Monoclonal | |
| 0.2mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.