missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD11c Chimeric Recombinant Mouse Monoclonal Antibody (N418)

Mouse Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA547950
This item is not returnable.
View return policy
Description
Specificity: The antibody binds to mouse and human CD11c.
CD11, along with CD18, form a heterodimer adhesion molecule. In particular, CD11 is composed of CD11a, CD11b and CD11c. CD11a is a leukocyte marker that is expressed in B and T lymphocytes, macrophages, monocytes, neutrophils, basophils and eosinophils. CD11b is primarily expressed by monocytes, macrophages, natural killer cells, some B and T-cells, and granulocytes. CD11c is expressed in monocytes, macrophages, natural killer cells, some granulocytes and less so in a subset of lymphocytes.
Specifications
| CD11c Chimeric | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.02% ProClin 300 | |
| P20702, Q9QXH4 | |
| Itgax | |
| Hamster spleen cells that had been primed to mouse spleen dendritic cells. | |
| 200 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 κ |
| Flow Cytometry, Immunocytochemistry | |
| N418 | |
| Unconjugated | |
| Itgax | |
| AI449405; CD11 antigen-like family member C; CD11c; CD11C (p150) alpha polypeptide; complement component 3 receptor 4 subunit; complement receptor 4; Cr4; integrin alpha X; integrin alpha-X; integrin aX; integrin subunit alpha X; integrin, alpha X; integrin, alpha X (antigen CD11C (p150), alpha polypeptide); integrin, alpha X (complement component 3 receptor 4 subunit); Itgax; Leu M5; leu M5, alpha subunit; leukocyte adhesion glycoprotein p150,95 alpha chain; Leukocyte adhesion receptor p150,95; leukocyte surface antigen p150,95, alpha subunit; myeloid membrane antigen, alpha subunit; N418; p150 95 integrin alpha chain; RGD1561123; SLEB6 | |
| Mouse | |
| Protein A | |
| RUO | |
| 16411, 3687 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction