missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD11 Antibody (24), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-2623F
This item is not returnable.
View return policy
Description
CD11 Monoclonal antibody specifically detects CD11 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunofluorescence
Specifications
| CD11 | |
| Monoclonal | |
| FITC | |
| antigen CD11b (p170), CD11 antigen-like family member B, CD11b, CD11b antigen, CD11B integrin, alpha M (complement component receptor 3, alpha; also known as CD11b(p170), macrophage antigen alpha polypeptide), Cell surface glycoprotein MAC-1 subunit alpha, CR-3 alpha chain, CR3AMGC117044, integrin alpha-M, integrin, alpha M (complement component 3 receptor 3 subunit), Leukocyte adhesion receptor MO1, MAC-1, MAC1A, macrophage antigen alpha polypeptide, MO1A, Neutrophil adherence receptor, neutrophil adherence receptor alpha-M subunit, SLEB6 | |
| Fibronectin-purified human monocytes . | |
| 0.1 mL | |
| Breast Cancer, Cancer, Immunology, Innate Immunity, Microglia Markers, Neuroscience, Signal Transduction | |
| 3684 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry, Immunofluorescence | |
| 24 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction