Learn More
Invitrogen™ CD102 (ICAM-2) Monoclonal Antibody (CBR-IC2/2), eBioscience™
Mouse Monoclonal Antibody
144.00 € - 1586.00 €
Specifications
| Antigen | CD102 (ICAM-2) |
|---|---|
| Clone | CBR-IC2/2 |
| Concentration | 0.5 mg/mL |
| Content And Storage | 4°C |
| Applications | Flow Cytometry, Functional Assay, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17130542
|
Invitrogen™
14-1029-37 |
2 mg |
1586.00 €
2mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15207147
|
Invitrogen™
14-1029-82 |
100 μg |
144.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Description: The CBRIC2/2 monoclonal antibody reacts with human ICAM-2 (CD102). ICAM-2 is a 55-65 kD transmembrane glycoprotein possessing 2 extracellular Ig domains, a single transmembrane domain, and a short 26-amino acid cytoplasmic domain. ICAM-2 is expressed on most leukocytes, and is strongly expressed on vascular endothelial cells. ICAM-2 is a ligand of integrin CD11a/CD18 (LFA-1), and also has been reported to bind integrin CD11b/CD18 (Mac-1). ICAM-2 is an intercellular adhesion molecule involved in lymphocyte homing to sites of inflammation. The CBRIC2/2 antibody has been reported to have blocking activity. Applications Reported: This CBRIC2/2 antibody has been reported for use in flow cytometric analysis, immunoblotting, immunoprecipitation, immunohistochemical staining, and functional assays. Applications Tested: This CBRIC2/2 antibody has been tested by flow cytometric analysis of human peripheral blood cells. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. The CBRIC2/2 antibody has also been found useful for immunoblotting of a human peripheral blood cell lysate, recognizing a protein of approximately 55 kD under non-reducing conditions. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. P...
ICAM-2 is a member of the intercellular adhesion molecule (ICAM) family. All ICAM proteins are type I transmembrane glycoproteins, contain 2-9 immunoglobulin-like C2-type domains, and bind to the leukocyte adhesion LFA-1 protein. ICAM-2 may play a role in lymphocyte recirculation by blocking LFA-1-dependent cell adhesion. It mediates adhesive interactions important for antigen-specific immune response, NK-cell mediated clearance, lymphocyte recirculation, and other cellular interactions important for immune response and surveillance. Several transcript variants encoding the same protein have been found for this gene. Diseases associated with ICAM2 include Colon Carcinoma In Situ.Specifications
| CD102 (ICAM-2) | |
| 0.5 mg/mL | |
| Flow Cytometry, Functional Assay, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| P13598 | |
| 3384 | |
| Primary | |
| Affinity chromatography |
| CBR-IC2/2 | |
| 4°C | |
| Monoclonal | |
| Liquid | |
| IgG2a | |
| Human | |
| ICAM2 | |
| CD102; Icam2; ICAM-2; Intercellular adhesion molecule 2; Ly-60; Lymphocyte function-associated AG-1 counter-receptor; RP23-418O11.10-001 | |
| ICAM2 | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.