Learn More
CCT251545, MedChemExpress
Product Code. 16465946
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16465946
Brand: Medchem Express HY12681
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress CCT251545 is an orally bioavailable and potent inhibitor of WNT signaling with an IC50 of 5 nM in 7dF3 cells. CCT251545 is a selective chemical probe for exploring the role of CDK8 and CDK19 in human disease.
CCT251545 is an orally bioavailable and potent inhibitor of WNT signaling with an IC50 of 5 nM in 7dF3 cells. CCT251545 is a selective chemical probe for exploring the role of CDK8 and CDK19 in human disease.
Specifications
Chemical Name or Material CCT251545
CAS 1661839-45-7
Color White
Purity Grade Notes Research
Molecular Formula C23H24ClN5O
Quantity 100 mg
Solubility Information DMSO : ≥ 50 mg/mL (118.51 mM)
SMILES O=C1NCCC12CCN(C3=C(Cl)C=NC=C3C4=CC=C(C5=CN(C)N=C5)C=C4)CC2
Molecular Weight (g/mol) 421.92
Formula Weight 421.92
Percent Purity 98.64%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease