missing translation for 'onlineSavingsMsg'
Learn More

CCT251545, MedChemExpress

Product Code. 16421497
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16421497

Brand: Medchem Express HY12681

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CCT251545 is an orally bioavailable and potent inhibitor of WNT signaling with an IC50 of 5 nM in 7dF3 cells. CCT251545 is a selective chemical probe for exploring the role of CDK8 and CDK19 in human disease.

CCT251545 is an orally bioavailable and potent inhibitor of WNT signaling with an IC50 of 5 nM in 7dF3 cells. CCT251545 is a selective chemical probe for exploring the role of CDK8 and CDK19 in human disease.

Specifications

Chemical Name or Material CCT251545
CAS 1661839-45-7
Color White
Purity Grade Notes Research
Molecular Formula C23H24ClN5O
Quantity 50 mg
Solubility Information DMSO : ≥ 50 mg/mL (118.51 mM)
SMILES O=C1NCCC12CCN(C3=C(Cl)C=NC=C3C4=CC=C(C5=CN(C)N=C5)C=C4)CC2
Molecular Weight (g/mol) 421.92
Formula Weight 421.92
Percent Purity 98.64%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.