Learn More
CCT241533 hydrochloride, MedChemExpress
Product Code. 16478737
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16478737
Brand: Medchem Express HY14715B
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress CCT241533 hydrochloride is a potent and selective CHK2 inhibitor with an IC50 of 3 nM and a Ki of 1.16 nM.
CCT241533 hydrochloride is a potent and selective CHK2 inhibitor with an IC50 of 3 nM and a Ki of 1.16 nM.
Specifications
Chemical Name or Material CCT241533 hydrochloride
CAS 1431697-96-9
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C23H28ClFN4O4
Quantity 5 mg
Solubility Information DMSO : ≥ 100 mg/mL (208.79 mM) ∣H2O : 33.33 mg/mL (69.59 mM; Need ultrasonic)
SMILES [H]Cl.COC1=C(OC)C=C(C(N[C@H]2[C@H](C(C)(C)O)CNC2)=NC(C3=C(O)C=CC(F)=C3)=N4)C4=C1
Molecular Weight (g/mol) 478.94
Formula Weight 478.94
Percent Purity 99.03%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease