missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CCSER2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557967
This item is not returnable.
View return policy
Description
Immunogen sequence: IQLLELQLAT QHICHQKCKE EKCTYADKYT QTPWRRIPGG YSAPSFSPWQ GSFQGIPRTV PPHRRQTSST TAFQQPSQTH RSHPGKTNKA TT Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 60%.
Specifications
| CCSER2 | |
| Polyclonal | |
| Unconjugated | |
| CCSER2 | |
| 1700012P13Rik; 2900054P12Rik; AI035535; AW108503; bA486O22.1; Ccser2; coiled-coil serine rich 2; coiled-coil serine rich protein 2; coiled-coil serine-rich protein 2; FAM190B; family with sequence similarity 190, member B; Gcap14; granule cell antiserum positive 14; granule cell antiserum positive protein 14; KIAA1128; NPD012; protein FAM190B; Protein GCAP14; Protein GCAP14 homolog; RGD1304626; serine-rich coiled-coil domain-containing protein 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 54462 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H7U1 | |
| CCSER2 | |
| Recombinant protein corresponding to Human CCSER2. Recombinant protein control fragment (Product #RP-97048). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction