Learn More
CCR6 Antibody (53103), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB195MFV450
566.10 EUR valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
Product Code. 18661520
  • 708.00 € / 0.10mL
Define Immunology Research on Your Terms With R&D Systems
Use promo code 24090
View all promotions
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
CCR6 Monoclonal antibody specifically detects CCR6 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CCR6
Monoclonal
mFluor Violet 450 SE
50mM Sodium Borate
Mouse
Protein A or G purified from ascites
RUO
Primary
Human
Purified
Flow Cytometry, Immunohistochemistry, CyTOF
53103
Flow Cytometry, Immunohistochemistry, CyTOF-ready
BN-1, CC-CKR-6, CCR-6, CD196 antigen, chemokine (C-C motif) receptor 6, CKR-L3G-protein coupled receptor 29, CKRL3LARC receptor, CMKBR6STRL22, DRY6, GPR29C-C CKR-6, GPRCY4DRY-6, lymphocyte, 22
NS0 mouse myeloma cell line transfected with human CCR6, Met1-Met374, Accession # P51684
0.1 mL
Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction
1235
Store at 4C in the dark.
IgG2b