missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CCP3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15742131
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15742131 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy

For Research Use Only. All usage must comply with product instructions.

Product Code. 15742131 Supplier Invitrogen™ Supplier No. PA554196

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For Research Use Only. All usage must comply with product instructions.

Rabbit Polyclonal Antibody

Immunogen sequence: CPEPVYIPTG LETQPLYPDS KEATVVYLAE DAYKEPCFVY SRVGGNRTPL KQPVDYRDNT LMFEARFESG NLQKVVKVAE YEY Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 86%.

Metallocarboxypeptidase that mediates both deglutamylation and deaspartylation of target proteins. Catalyzes the deglutamylation of polyglutamate side chains generated by post-translational polyglutamylation in proteins such as tubulins. Also removes gene-encoded polyglutamates or polyaspartates from the carboxy-terminus of target proteins such as MYLK. Does not show detyrosinase or deglycylase activities from the carboxy-terminus of tubulin. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCP3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Agbl3
Gene Accession No. Q8NEM8
Gene Alias 2900053G10Rik; 4930431N21Rik; 6530406M24Rik; Agbl3; ATP/GTP binding protein like 3; ATP/GTP binding protein-like 3; ATP/GTP-binding protein-like 3; CBCP3; CCP3; Cytosolic carboxypeptidase 3; RGD1561995
Gene Symbols Agbl3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CCP3. Recombinant protein control fragment (Product #RP-92759).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 340351
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.