missing translation for 'onlineSavingsMsg'
Learn More

CCL15, Rabbit anti-Human, Polyclonal Antibody, Abnova™

Product Code. 16132618
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16132618 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16132618 Supplier Abnova Supplier No. H00006359D01.100uL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Sequence: MKVSVAALSCLMLVAVLGSQAQFINDAETELMMSKLPLENPVVLNSFHFAADCCTSYISQSIPCSLMKSYFETSSECSKPGVIFLTKKGRQVCAKPSGPGVQDCMKKLKPYSI

Specifications

Antigen CCL15
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation No additive
Gene chemokine (C-C motif) ligand 15
Gene Accession No. NM_004167
Gene Alias HCC-2/HMRP-2B/LKN1/Lkn-1/MIP-1d/MIP-5/NCC-3/NCC3/SCYA15/SCYL3/SY15
Gene Symbols CCL15
Host Species Rabbit
Immunogen CCL15 (NP_004158.2, 1 a.a. ∼ 113 a.a) full-length human protein.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6359
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.