Learn More
CCL11/Eotaxin Rat anti-Mouse, Clone: 42285, R&D Systems™

Rat Monoclonal Antibody has been used in 5 publications
Brand: R&D Systems MAB420-SP
Description
CCL11/Eotaxin Monoclonal specifically detects CCL11/Eotaxin in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Neutralization.Specifications
| CCL11/Eotaxin | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| C-C motif chemokine 11, chemokine (C-C motif) ligand 11, Eosinophil chemotactic protein, Eotaxin, eotaxin-1, MGC22554, SCYA11Small-inducible cytokine A11, small inducible cytokine subfamily A (Cys-Cys), member 11 (eotaxin) | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6356 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot, Immunohistochemistry, Neutralization | |
| 42285 | |
| Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Neutralization 3-30 ug/mL | |
| P48298 | |
| CCL11 | |
| E. coli-derived recombinant mouse CCL11/Eotaxin His24-Pro97 Accession # P48298 | |
| 25 μg | |
| Primary | |
| Detects mouse CCL11/Eotaxin in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 10% cross-reactivity with recombinant human (rh) CCL11/Eotaxin, recombinant mouse CCL7, and rhCCL21 is observed. | |
| Mouse | |
| Purified |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.