Learn More
CCG215022, MedChemExpress
Product Code. 16487678
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
Product Code. 16487678
Brand: Medchem Express HY18991
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress CCG215022 is a G protein-coupled receptor kinases (GRKs) inhibitor with IC50s of 0.15±0.07 μM, 0.38±0.06 μM and 3.9±1 μM for GRK2, GRK5 and GRK1, respectively.
CCG215022 is a G protein-coupled receptor kinases (GRKs) inhibitor with IC50s of 0.15±0.07 μM, 0.38±0.06 μM and 3.9±1 μM for GRK2, GRK5 and GRK1, respectively.
Specifications
Chemical Name or Material CCG215022
CAS 1813527-81-9
Color Off-White
Purity Grade Notes Research
Molecular Formula C26H22FN7O3
Quantity 10 mg
Solubility Information DMSO : ≥ 28 mg/mL (56.06 mM)
SMILES CC(N1)=C(C(NC2=CC=C(NN=C3)C3=C2)=O)C(C4=CC(C(NCC5=CC=CC=N5)=O)=C(F)C=C4)NC1=O
Molecular Weight (g/mol) 499.5
Formula Weight 499.5
Percent Purity 98.03%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease