missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CCDC94 Polyclonal antibody specifically detects CCDC94 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | CCDC94 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | DyLight 350 |
| Dilution | Western Blot |
| Formulation | 50mM Sodium Borate |
| Gene Alias | coiled-coil domain containing 94, coiled-coil domain-containing protein 94, FLJ10374 |
| Host Species | Rabbit |
| Immunogen | Recognizes region between residues Between between 273 and 323.. |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?