missing translation for 'onlineSavingsMsg'
Learn More

CC-401 hydrochloride, MedChemExpress

Product Code. 16441667
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16441667

Brand: Medchem Express HY13022

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CC-401 hydrochloride is a potent inhibitor of all three forms of JNK with Ki of 25 to 50 nM.

CC-401 hydrochloride is a potent inhibitor of all three forms of JNK with Ki of 25 to 50 nM.

Specifications

Chemical Name or Material CC-401 hydrochloride
CAS 1438391-30-0
Color Off-White
Purity Grade Notes Research
Molecular Formula C22H25ClN6O
Quantity 50 mg
Synonym CC401 HCl
Solubility Information DMSO : 100 mg/mL (235.33 mM; Need ultrasonic) ∣H2O : 12.5 mg/mL (29.42 mM; Need ultrasonic)
SMILES C1(C2=CC=CC(OCCN3CCCCC3)=C2)=NNC4=C1C=C(C=C4)C5=NC=NN5.Cl
Molecular Weight (g/mol) 424.93
Formula Weight 424.93
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.