missing translation for 'onlineSavingsMsg'
Learn More

CC-401 hydrochloride, MedChemExpress

Product Code. p-200101858
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16485488 10 mg 10mg
16441667 50 mg 50mg
16428207 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16485488 Supplier Medchem Express Supplier No. HY13022
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress CC-401 hydrochloride is a potent inhibitor of all three forms of JNK with Ki of 25 to 50 nM.

CC-401 hydrochloride is a potent inhibitor of all three forms of JNK with Ki of 25 to 50 nM.

Specifications

Chemical Name or Material CC-401 hydrochloride
CAS 1438391-30-0
Color Off-White
Purity Grade Notes Research
Molecular Formula C22H25ClN6O
Quantity 10 mg
Synonym CC401 HCl
Solubility Information DMSO : 100 mg/mL (235.33 mM; Need ultrasonic) ∣H2O : 12.5 mg/mL (29.42 mM; Need ultrasonic)
SMILES C1(C2=CC=CC(OCCN3CCCCC3)=C2)=NNC4=C1C=C(C=C4)C5=NC=NN5.Cl
Molecular Weight (g/mol) 424.93
Formula Weight 424.93
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.