Learn More
CC-401 hydrochloride, MedChemExpress
Product Code. 16485488
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16485488 10 mg 10mg
16441667 50 mg 50mg
16428207 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16485488 Supplier Medchem Express Supplier No. HY13022
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
MedChemExpress CC-401 hydrochloride is a potent inhibitor of all three forms of JNK with Ki of 25 to 50 nM.
CC-401 hydrochloride is a potent inhibitor of all three forms of JNK with Ki of 25 to 50 nM.
Specifications
Chemical Name or Material CC-401 hydrochloride
CAS 1438391-30-0
Color Off-White
Purity Grade Notes Research
Molecular Formula C22H25ClN6O
Quantity 10 mg
Synonym CC401 HCl
Solubility Information DMSO : 100 mg/mL (235.33 mM; Need ultrasonic) ∣H2O : 12.5 mg/mL (29.42 mM; Need ultrasonic)
SMILES C1(C2=CC=CC(OCCN3CCCCC3)=C2)=NNC4=C1C=C(C=C4)C5=NC=NN5.Cl
Molecular Weight (g/mol) 424.93
Formula Weight 424.93
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease