missing translation for 'onlineSavingsMsg'
Learn More

CC-115 hydrochloride, MedChemExpress

Product Code. p-200097584
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16427228

Brand: Medchem Express HY16962A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CC-115 hydrochloride is a potent and dual DNA-PK and mTOR kinase inhibitor with IC50s of 13 nM and 21 nM, respectively. CC-115 blocks both mTORC1 and mTORC2 signaling.

CC-115 hydrochloride is a potent and dual DNA-PK and mTOR kinase inhibitor with IC50s of 13 nM and 21 nM, respectively. CC-115 blocks both mTORC1 and mTORC2 signaling.

Specifications

Chemical Name or Material CC-115 hydrochloride
CAS 1300118-55-1
Color Yellow
Purity Grade Notes Research
Molecular Formula C16H17ClN8O
Quantity 10 mg
Solubility Information H2O : 50 mg/mL (134.12 mM; Need ultrasonic) ∣DMSO : ≥ 30 mg/mL (80.47 mM)
SMILES [H]Cl.O=C1CN=C2C(N1CC)=NC(C3=C(C)N=C(C4=NN=CN4)C=C3)=CN2
Molecular Weight (g/mol) 372.81
Formula Weight 372.81
Percent Purity 98.23%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.