Learn More
CC-115 hydrochloride, MedChemExpress
Product Code. 16427228
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Product Code. 16427228
Brand: Medchem Express HY16962A
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress CC-115 hydrochloride is a potent and dual DNA-PK and mTOR kinase inhibitor with IC50s of 13 nM and 21 nM, respectively. CC-115 blocks both mTORC1 and mTORC2 signaling.
CC-115 hydrochloride is a potent and dual DNA-PK and mTOR kinase inhibitor with IC50s of 13 nM and 21 nM, respectively. CC-115 blocks both mTORC1 and mTORC2 signaling.
Specifications
Chemical Name or Material CC-115 hydrochloride
CAS 1300118-55-1
Color Yellow
Purity Grade Notes Research
Molecular Formula C16H17ClN8O
Quantity 10 mg
Solubility Information H2O : 50 mg/mL (134.12 mM; Need ultrasonic) ∣DMSO : ≥ 30 mg/mL (80.47 mM)
SMILES [H]Cl.O=C1CN=C2C(N1CC)=NC(C3=C(C)N=C(C4=NN=CN4)C=C3)=CN2
Molecular Weight (g/mol) 372.81
Formula Weight 372.81
Percent Purity 98.23%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease