missing translation for 'onlineSavingsMsg'
Learn More

CBB1007 trihydrochloride, MedChemExpress

Product Code. 16482547
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16482547 50 mg 50mg
16446448 10 mg 10mg
16419037 5 mg 5mg
16406736 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16482547 Supplier Medchem Express Supplier No. HY15313C
 Restricted Item
Estimated Shipment: 31-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress CBB1007 trihydrochloride is a cell-permeable amidino-guanidinium compound that acts as a potent, reversible and substrate competitive LSD1 selective inhibitor (IC50 = 5.27 (mu)M for hLSD1).

CBB1007 trihydrochloride is a cell-permeable amidino-guanidinium compound that acts as a potent, reversible and substrate competitive LSD1 selective inhibitor (IC50 = 5.27 (mu)M for hLSD1).

Specifications

Chemical Name or Material CBB1007 trihydrochloride
Color White
Purity Grade Notes Research
Molecular Formula C27H37Cl3N8O4
Quantity 50 mg
Solubility Information DMSO : ≥ 54 mg/mL (83.85 mM)
SMILES NC(N1CCN(CC1)CC2=CC(C(OC)=O)=CC(C(N3CCN(CC3)C(C4=CC=C(C=C4)C(N)=N)=O)=O)=C2)=N.[H]Cl.[H]Cl.[H]Cl
Molecular Weight (g/mol) 643.99
Formula Weight 643.99
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.