Learn More
CB 300919, MedChemExpress
Product Code. 16498647
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16498647 5 mg 5mg
16436048 10 mg 10mg
16432187 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16498647 Supplier Medchem Express Supplier No. HY14375
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
MedChemExpress CB 300919 is a quinazoline-based antitumour agent with high activity in the CH1 human ovarian tumour xenograft. CB 300919 has a continuous exposure (96 h) growth inhibition IC50 value of 2 nM in human CH1 ovarian tumor xenograft.
CB 300919 is a quinazoline-based antitumour agent with high activity in the CH1 human ovarian tumour xenograft. CB 300919 has a continuous exposure (96 h) growth inhibition IC50 value of 2 nM in human CH1 ovarian tumor xenograft.
Specifications
Chemical Name or Material CB 300919
Physical Form Solid
CAS 289715-28-2
Color Off-White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C32H34ClN7O2
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : ≥ 100 mg/mL (171.20 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES CN1CCN(CC1)CC2=NC3=C(C(N2C)=O)C=C(CN(CC#C)C4=CC=C(C=C4)C(NCC5=CC=CN=C5)=O)C(Cl)=C3
Molecular Weight (g/mol) 584.11
Formula Weight 584.11
Percent Purity 98.01%
Grade Research