Learn More
CAY10566, MedChemExpress
Product Code. 16476848
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
Unit Size:
10mg
1mg
25mg
5mg
Product Code. 16476848
Brand: Medchem Express HY15823
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress CAY10566 is a potent, orally bioavailable and selective stearoyl-CoA desaturase1 (SCD1) inhibitor with IC50s of 4.5 and 26 nM in mouse and human enzymatic assays, respectively. CAY10566 also shows excellent cellular activity in blocking the conversion of saturated long-chain fatty acid-CoAs (LCFA-CoAs) to monounsaturated LCFA-CoAs in HepG2 cells (IC50=7.9 nM or 6.8 nM).
CAY10566 is a potent, orally bioavailable and selective stearoyl-CoA desaturase1 (SCD1) inhibitor with IC50s of 4.5 and 26 nM in mouse and human enzymatic assays, respectively.
Specifications
Chemical Name or Material CAY10566
CAS 944808-88-2
Color Yellow
Purity Grade Notes Research
Molecular Formula C18H17ClFN5O2
Quantity 10 mg
Solubility Information DMSO : 25 mg/mL (64.13 mM; Need ultrasonic)
SMILES CC1=NN=C(C2=NN=C(N3CCC(OC4=CC(F)=CC=C4Cl)CC3)C=C2)O1
Molecular Weight (g/mol) 389.81
Formula Weight 389.81
Percent Purity 98.07%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death